accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 599-71-3

IUPAC Name: N-hydroxybenzenesulfonamide
Synonyms: benzenesulfonamide, N-hydroxy-
Molecular Weight (g/mol): 173.19
Molecular Formula: C6H7NO3S
InChi Key: BRMDATNYMUMZLN-UHFFFAOYSA-N
SMILES: ONS(=O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)