Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Hydroxybenzenesulfonamide 97%, Thermo Scientific™

Catalog No. AC411940250
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC411940250 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC411940250 Supplier Thermo Scientific Chemicals Supplier No. 411940250

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 599-71-3
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Molecular Formula C6H7NO3S
Quantity 25g
Synonym piloty's acid, benzenesulfohydroxamic acid, benzenesulfonamide, n-hydroxy, benzenesulphonohydroxamic acid, benzsulfohydroxamic acid, hydroxamic acid, benzsulfo, n-phenylsulfonyl hydroxylamine, hydroxylamine, n-phenylsulfonyl, unii-c64l4umh19, n-hydroxy-benzenesulfonamide
InChI Key BRMDATNYMUMZLN-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)S(=O)(=O)NO
IUPAC Name N-hydroxybenzenesulfonamide
Molecular Weight (g/mol) 173.186
PubChem CID 69033
Formula Weight 173.19
Percent Purity ≥96.0%
Chemical Name or Material N-Hydroxybenzenesulfonamide, 97%
Show More Show Less
EINECSNumber 209-973-8
RTECSNumber MX9350000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.