accessibility menu, dialog, popup

UserName

Viewing profile as user:
POH2N

CAS RN 5994-87-6

IUPAC Name: [amino(phenyl)phosphoroso]benzene
Synonyms: [amino(phenyl)phosphoroso]benzene
Molecular Weight (g/mol): 217.21
Molecular Formula: C12H12NOP
InChi Key: RIGIWEGXTTUCIQ-UHFFFAOYSA-N
SMILES: NP(=O)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)