accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOO

CAS RN 59985-82-9

IUPAC Name: 5-phenyl-1,2-oxazole-3-carbaldehyde
Synonyms: 5-phenyl-1,2-oxazole-3-carbaldehyde
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: QRHAQXZGSWFBOK-UHFFFAOYSA-N
SMILES: O=CC1=NOC(=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results