CAS RN 60-18-4
Thermo Scientific Chemicals L-Tyrosine, 99%
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine [Free Base], MP Biomedicals
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine, MP Biomedicals™
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Thermo Scientific Chemicals L-Tyrosine, Cell Culture Reagent
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Tyrosine, USP, 98.5-101.5%, Spectrum™ Chemical
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine, FCC, 98.5-101.5%, Spectrum™ Chemical
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
L-Tyrosine, Pharmaceutical Secondary Standard, Certified Reference Material, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
L-(-)-Tyrosine 98.5+%, TCI America™
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N Synonym: l-tyrosine,tyrosine,s-tyrosine,p-tyrosine,h-tyr-oh,l-p-tyrosine,2s-2-amino-3-4-hydroxyphenyl propanoic acid,4-hydroxy-l-phenylalanine,l---tyrosine,tyrosine, l PubChem CID: 6057 ChEBI: CHEBI:17895 IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
H-Tyr-OH, Advanced ChemTech
CAS: 60-18-4 Molecular Formula: C9H11NO3 Molecular Weight (g/mol): 181.19 MDL Number: MFCD00002606 InChI Key: OUYCCCASQSFEME-SVGMAFHSNA-N IUPAC Name: (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid SMILES: N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Medisca Tyrosine (L), USP (Dietary Supplement Grade)
Tyrosine (L), USP (Dietary Supplement Grade) comes as white, odorless crystals or crystalline powder. Very slightly soluble in water and insoluble in alcohol.
Thermo Scientific Production Chemicals and Services L-Tyrosine USP/EP/FCC, Ajinomoto
CAS: 60-18-4 Molecular Weight (g/mol): 181.19
Thermo Scientific Production Chemicals and Services L-Tyrosine USP/EP/FCC, Ajinomoto
CAS: 60-18-4 Molecular Weight (g/mol): 181.19
Thermo Scientific Production Chemicals and Services L-Tyrosine USP/EP/FCC, Ajinomoto
CAS: 60-18-4 Molecular Weight (g/mol): 181.19
- 1
- 2