Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18607629 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-629 50 kg Fiber Drum
18-607-625 100 g Poly Bottle
18-607-626 1 kg Poly Bottle
18-607-627 5 kg Poly Pail
18-607-628 25 kg Fiber Drum
18-607-624 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-607-629 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T115350KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T1153, 60-18-4, C9H11NO3

Spectrum™ Chemical Tyrosine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.004
Loss on Drying 0.003
Packaging Fiber Drum
Specific Rotation -9.8° to -11.2°
Quantity 50 kg
Percent Purity 98.5 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.