accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCH3OONCH3CH3H3C

CAS RN 60-31-1

IUPAC Name: [2-(acetyloxy)ethyl]trimethylazanium chloride
Synonyms: acetylcholine chloride 氯乙酰胆碱 chloroacetylcholine miochol
Molecular Weight (g/mol): 181.66
Molecular Formula: C7H16ClNO2
InChi Key: JUGOREOARAHOCO-UHFFFAOYSA-M
SMILES: [Cl-].CC(=O)OCC[N+](C)(C)C
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (7)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (7)
  • (1)