CAS RN 60-33-3
Linoleic acid, 95%
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.452 MDL Number: MFCD00064241 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Thermo Scientific Chemicals Linoleic acid, 70%
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.45 MDL Number: MFCD00064241 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Linoleic acid, tech. 75%
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.452 MDL Number: MFCD00064241 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Thermo Scientific Chemicals Linoleic acid, 99%
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.45 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Linoleic Acid 97.0+%, TCI America™
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.452 MDL Number: MFCD00064241 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Linoleic Acid 85.0+%, TCI America™
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.452 MDL Number: MFCD00064241 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N Synonym: linoleic acid,linolic acid,telfairic acid,cis,cis-linoleic acid,9z,12z-octadeca-9,12-dienoic acid,linoleate,grape seed oil,cis,cis-9,12-octadecadienoic acid,cis-9,cis-12-octadecadienoic acid,9z,12z-linoleic acid PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Agilent Technologies Linoleic Acid, 99+%, Ultra Scientific
CAS: 60-33-3 Molecular Formula: C18H32O2 Molecular Weight (g/mol): 280.452 InChI Key: OYHQOLUKZRVURQ-HZJYTTRNSA-N PubChem CID: 5280450 ChEBI: CHEBI:17351 IUPAC Name: (9Z,12Z)-octadeca-9,12-dienoic acid SMILES: CCCCCC=CCC=CCCCCCCCC(=O)O
Sigma Aldrich Linoleic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.