Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Linoleic Acid, 99+%, Ultra Scientific

Catalog No. USFLSA089
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-FLSA-089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-FLSA-089 Supplier Agilent Technologies Supplier No. FLSA089
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60-33-3
Molecular Formula C18H32O2
Molecular Weight (g/mol) 280.452
InChI Key OYHQOLUKZRVURQ-HZJYTTRNSA-N
PubChem CID 5280450
ChEBI CHEBI:17351
IUPAC Name (9Z,12Z)-octadeca-9,12-dienoic acid
SMILES CCCCCC=CCC=CCCCCCCCC(=O)O

Specifications

CAS Max % 100.0
Assay Percent Range >99%
Physical Form Liquid
Quantity 100 mg
Chemical Name or Material Linoleic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.