accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNOSPSOH3COH3C

CAS RN 60-51-5

IUPAC Name: O,O-dimethyl {[(methylcarbamoyl)methyl]sulfanyl}phosphonothioate
Synonyms: dimethoate phosphamide
Molecular Weight (g/mol): 229.25
Molecular Formula: C5H12NO3PS2
InChi Key: MCWXGJITAZMZEV-UHFFFAOYSA-N
SMILES: CNC(=O)CSP(=S)(OC)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results