CAS RN 600-18-0
IUPAC Name:
2-oxobutanoic acid
Synonyms:
2-oxobutanoic acid
Molecular Weight (g/mol):
102.09
Molecular Formula:
C4H6O3
InChi Key:
TYEYBOSBBBHJIV-UHFFFAOYSA-N
SMILES:
CCC(=O)C(O)=O
1
–
1
of
1
results
1
2-Oxobutyric Acid 98.0+%, TCI America™
CAS: 600-18-0 Molecular Formula: C4H6O3 Molecular Weight (g/mol): 102.09 MDL Number: MFCD00004164 InChI Key: TYEYBOSBBBHJIV-UHFFFAOYSA-N PubChem CID: 58 ChEBI: CHEBI:30831 IUPAC Name: 2-oxobutanoic acid SMILES: CCC(=O)C(O)=O