Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Oxobutyric Acid 98.0+%, TCI America™
SDP

Catalog No. O03645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O03645G Supplier TCI America Supplier No. O03645G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 600-18-0
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00004164
InChI Key TYEYBOSBBBHJIV-UHFFFAOYSA-N
PubChem CID 58
ChEBI CHEBI:30831
IUPAC Name 2-oxobutanoic acid
SMILES CCC(=O)C(O)=O

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 84°C
Quantity 5 g
Formula Weight 102.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Oxobutyric Acid
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.