CAS RN 6004-24-6
(1-Hexadecyl)pyridinium chloride monohydrate, 98%
CAS: 6004-24-6 Molecular Formula: C21H40ClNO Molecular Weight (g/mol): 358.01 MDL Number: MFCD00149977 InChI Key: NFCRBQADEGXVDL-UHFFFAOYSA-M Synonym: cetylpyridinium chloride monohydrate,1-hexadecylpyridin-1-ium chloride hydrate,1-hexadecylpyridinium chloride monohydrate,hexadecylpyridinium chloride monohydrate,unii-d9om4sk49p,pyridinium, 1-hexadecyl-, chloride, monohydrate,d9om4sk49p,cetylpyridinii chloridum,sprol tn PubChem CID: 22324 ChEBI: CHEBI:3566 SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Cetylpyridinium Chloride, Monohydrate, USP, 98-102%, Spectrum™ Chemical
CAS: 6004-24-6 Molecular Formula: C21H40ClNO Molecular Weight (g/mol): 358.01 MDL Number: MFCD00149977 InChI Key: NFCRBQADEGXVDL-UHFFFAOYSA-M IUPAC Name: 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
1-Hexadecylpyridinium chloride monohydrate, 96.0-101.0%
CAS: 6004-24-6 Molecular Formula: C21H40ClNO Molecular Weight (g/mol): 358.01 MDL Number: MFCD00149977 InChI Key: NFCRBQADEGXVDL-UHFFFAOYSA-M Synonym: cetylpyridinium chloride monohydrate,1-hexadecylpyridin-1-ium chloride hydrate,1-hexadecylpyridinium chloride monohydrate,hexadecylpyridinium chloride monohydrate,unii-d9om4sk49p,pyridinium, 1-hexadecyl-, chloride, monohydrate,d9om4sk49p,cetylpyridinii chloridum,sprol tn PubChem CID: 22324 ChEBI: CHEBI:3566 IUPAC Name: 1-hexadecylpyridin-1-ium;chloride;hydrate SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
CPC Monohydrate (=Cetylpyridinium Chloride Monohydrate) 98.0+%, TCI America™
CAS: 6004-24-6 Molecular Formula: C21H40ClNO Molecular Weight (g/mol): 358.01 MDL Number: MFCD00149977 InChI Key: NFCRBQADEGXVDL-UHFFFAOYSA-M Synonym: cetylpyridinium chloride monohydrate,1-hexadecylpyridin-1-ium chloride hydrate,1-hexadecylpyridinium chloride monohydrate,hexadecylpyridinium chloride monohydrate,unii-d9om4sk49p,pyridinium, 1-hexadecyl-, chloride, monohydrate,d9om4sk49p,cetylpyridinii chloridum,sprol tn PubChem CID: 22324 ChEBI: CHEBI:3566 IUPAC Name: 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Hexadecylpyridinium Chloride Monohydrate 98.0+%, TCI America™
CAS: 6004-24-6 Molecular Formula: C21H40ClNO Molecular Weight (g/mol): 358.01 MDL Number: MFCD00149977 InChI Key: NFCRBQADEGXVDL-UHFFFAOYSA-M Synonym: cetylpyridinium chloride monohydrate,1-hexadecylpyridin-1-ium chloride hydrate,1-hexadecylpyridinium chloride monohydrate,hexadecylpyridinium chloride monohydrate,unii-d9om4sk49p,pyridinium, 1-hexadecyl-, chloride, monohydrate,d9om4sk49p,cetylpyridinii chloridum,sprol tn PubChem CID: 22324 ChEBI: CHEBI:3566 IUPAC Name: 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Cetylpyridinium Chloride, G-Biosciences
CAS: 6004-24-6 Molecular Weight (g/mol): 358.00 g/mol