Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cetylpyridinium Chloride, G-Biosciences
SCI_ED

Catalog No. S100904
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S100904 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S100904 Supplier G-Biosciences Supplier No. RC962
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cetylpyridinium chloride is a cationic quaternary ammonium compound that has been used to eliminate oral pathogens. It has shown a high degree of efficacy towards gram-positive, gram-negative bacteria and some pathogenic fungi.

Cetylpyridinium chloride functions as a surfactant, reducing the surface tension between pathogenic membranes and their environments. Greater interaction between the membrane and environment disrupts the cell membrane and osmoregulation. Compromising the membrane ultimately results in cell death.

Chemical Identifiers

CAS 6004-24-6
Molecular Weight (g/mol) 358.00 g/mol

Specifications

Physical Form Flakes or Powder
Quantity 100 g
Percent Purity >98%
Grade Laboratory
Chemical Name or Material Cetylpyridinium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.