accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C

CAS RN 6006-33-3

IUPAC Name: tridecylcyclohexane
Synonyms: N-tridecylcyclohexane
Molecular Weight (g/mol): 266.51
Molecular Formula: C19H38
InChi Key: DKRUJKKWJGNYQN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results