Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecylcyclohexane 98.0+%, TCI America™
SDP

Catalog No. T076725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T076725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T076725ML Supplier TCI America Supplier No. T076725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6006-33-3
Molecular Formula C19H38
Molecular Weight (g/mol) 266.513
MDL Number MFCD00045507
InChI Key DKRUJKKWJGNYQN-UHFFFAOYSA-N
Synonym 1-Cyclohexyltridecane
PubChem CID 22327
IUPAC Name tridecylcyclohexane
SMILES CCCCCCCCCCCCCC1CCCCC1

Specifications

Color Colorless
Boiling Point 346°C
Quantity 25 mL
Formula Weight 266.51
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.