accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6009-98-9

IUPAC Name: sodium 2-(4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate
Synonyms: sodium 2-(4-{4,7-dihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethanesulfonate
Molecular Weight (g/mol): 521.69
Molecular Formula: C26H44NNaO6S
InChi Key: IYPNVUSIMGAJFC-UHFFFAOYNA-M
SMILES: [Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)