Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurochenodeoxycholic Acid Sodium Salt, Spectrum™ Chemical
SDP

Catalog No. 18607232 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-232 1 g Amber Glass Bottle
18-607-231 250 mg Amber Glass Bottle
18-607-230 100 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-232 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T39251GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T3925, 6009-98-9, C26H44NNaO6S

Spectrum™ Chemical Taurochenodeoxycholic Acid Sodium Salt , also known as 12-Deoxycholyltaurine or 12-Desoxycholyltaurine, is a bile acid that acts as detergent in the small intestine to solubilize fats. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 6009-98-9
Molecular Formula C26H44NNaO6S
Molecular Weight (g/mol) 521.69
InChI Key IYPNVUSIMGAJFC-UHFFFAOYNA-M
IUPAC Name sodium 2-(4-{4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl}pentanamido)ethane-1-sulfonate
SMILES [Na+].CC(CCC(=O)NCCS([O-])(=O)=O)C1CCC2C3C(O)CC4CC(O)CCC4(C)C3CCC12C

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.