accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OHOOHO

CAS RN 601-79-6

IUPAC Name: 2-(propan-2-yl)propanedioic acid
Synonyms: 2-isopropylpropanedioic acid
Molecular Weight (g/mol): 146.14
Molecular Formula: C6H10O4
InChi Key: DQEUFPARIOFOAI-UHFFFAOYSA-N
SMILES: CC(C)C(C(O)=O)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)