Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isopropylmalonic Acid 99.0+%, TCI America™
SDP

Catalog No. I05575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I05575G Supplier TCI America Supplier No. I05575G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 601-79-6
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
MDL Number MFCD00021717
InChI Key DQEUFPARIOFOAI-UHFFFAOYSA-N
Synonym isopropylmalonic acid, 2-isopropylmalonic acid, malonic acid, isopropyl, 2-isopropylpropanedioic acid, propanedioic acid, 1-methylethyl, .alpha.-carboxyisovaleric acid, 2-propan-2-yl propanedioic acid, isopropylmalonsaure, isopropyl malonic acid, iso-propyl malonic acid
PubChem CID 69037
IUPAC Name 2-(propan-2-yl)propanedioic acid
SMILES CC(C)C(C(O)=O)C(O)=O

Specifications

Melting Point 85°C
Color White
Quantity 5 g
UN Number 2811
Formula Weight 146.14
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Isopropylmalonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.