accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60111-54-8

H3CSiCH3OSiOSiH3CCH3H2COSiH3CCH3CH2OSiCH3H3CH2CCH2

CAS RN 60111-54-8

IUPAC Name: tetraethenyldimethylsilyl silicate
Synonyms: tetraethenyldimethylsilyl silicate
Molecular Weight (g/mol): 432.89
Molecular Formula: C16H36O4Si5
InChi Key: JMTMWSZLPHDWBQ-UHFFFAOYSA-N
SMILES: C[Si](C)(O[Si](O[Si](C)(C)C=C)(O[Si](C)(C)C=C)O[Si](C)(C)C=C)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results