Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis[dimethyl(vinyl)silyl] Orthosilicate 98.0+%, TCI America™
SDP

Catalog No. T35175ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35175ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T35175ML Supplier TCI America Supplier No. T35175ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60111-54-8
Molecular Formula C16H36O4Si5
Molecular Weight (g/mol) 432.89
MDL Number MFCD00271023
InChI Key JMTMWSZLPHDWBQ-UHFFFAOYSA-N
Synonym Tetrakis[dimethyl(vinyl)silyloxy]silane
PubChem CID 6453911
IUPAC Name tetraethenyldimethylsilyl silicate
SMILES C[Si](C)(O[Si](O[Si](C)(C)C=C)(O[Si](C)(C)C=C)O[Si](C)(C)C=C)C=C

Specifications

Color Colorless
Boiling Point 130°C
Quantity 5 mL
Formula Weight 432.89
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrakis[dimethyl(vinyl)silyl] Orthosilicate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.