accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60117-35-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 5-azido-2-nitrobenzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 5-azido-2-nitrobenzoate
Molecular Weight (g/mol): 305.21
Molecular Formula: C11H7N5O6
InChi Key: FUOJEDZPVVDXHI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1C(=O)ON1C(=O)CCC1=O)N=[N+]=[N-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results