Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Succinimidyl 5-Azido-2-nitrobenzoate 97.0+%, TCI America™
SDP

Catalog No. S086010MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S086010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S086010MG Supplier TCI America Supplier No. S086010MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 60117-35-3
Molecular Formula C11H7N5O6
Molecular Weight (g/mol) 305.206
MDL Number MFCD00054962
InChI Key FUOJEDZPVVDXHI-UHFFFAOYSA-N
Synonym N-(5-Azido-2-nitrobenzoyloxy)succinimide, 5-Azido-2-nitrobenzoic Acid N-Succinimidyl Ester, ANB-NOS
PubChem CID 3080866
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 5-azido-2-nitrobenzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=C(C=CC(=C2)N=[N+]=[N-])[N+](=O)[O-]

Specifications

Melting Point 136°C
Color Yellow
Quantity 10 mg
Formula Weight 305.21
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 5-Azido-2-nitrobenzoate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.