accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSClClCl

CAS RN 6012-97-1

IUPAC Name: tetrachlorothiophene
Synonyms: tetrachlorothiophene
Molecular Weight (g/mol): 221.9
Molecular Formula: C4Cl4S
InChi Key: WZXXZHONLFRKGG-UHFFFAOYSA-N
SMILES: ClC1=C(Cl)C(Cl)=C(Cl)S1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)