Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrachlorothiophene, 98%, Thermo Scientific™

Catalog No. AAL0312706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0312706 5 g
AAL0312703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0312706 Supplier Thermo Scientific Chemicals Supplier No. L0312706

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6012-97-1
Molecular Formula C4Cl4S
Molecular Weight (g/mol) 221.904
MDL Number MFCD00005422
InChI Key WZXXZHONLFRKGG-UHFFFAOYSA-N
Synonym tetrachlorothiophene, perchlorothiophene, thiophene, tetrachloro, pemphene, penphene, tetrachlorothiofene, if fumigant, tctp, penn salt td-183, tetrachlorthiofen
PubChem CID 22344
IUPAC Name 2,3,4,5-tetrachlorothiophene
SMILES C1(=C(SC(=C1Cl)Cl)Cl)Cl

Specifications

Melting Point 26°C to 30°C
Density 1.7
Boiling Point 232°C to 233°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.591
Quantity 5 g
UN Number UN2811
Beilstein 118374
Formula Weight 221.92
Percent Purity 98%
Chemical Name or Material Tetrachlorothiophene
Show More Show Less
Hazard Category H301+H311+H331
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 227-866-4
RTECSNumber XN0350000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.