accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)(R)OOHOHOCH3

CAS RN 60134-26-1

IUPAC Name: (2R,3S)-2-(hydroxymethyl)-5-methoxyoxolan-3-ol
Synonyms: (2R,3S)-2-(hydroxymethyl)-5-methoxyoxolan-3-ol
Molecular Weight (g/mol): 148.16
Molecular Formula: C6H12O4
InChi Key: NVGJZDFWPSOTHM-XCSHRFQENA-N
SMILES: COC1C[C@H](O)[C@@H](CO)O1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)