accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClClClClCl(?)

CAS RN 60145-22-4

IUPAC Name: 2,2',4,4',5,6'-hexachloro-1,1'-biphenyl
Synonyms: 2,2',4,4',5,6'-hexachloro-1,1'-biphenyl
Molecular Weight (g/mol): 360.86
Molecular Formula: C12H4Cl6
InChi Key: QXZHEJWDLVUFFB-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(C(Cl)=C1)C1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results