accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPH3COOCH3

CAS RN 60146-72-7

IUPAC Name: bis(2-methylphenyl) methylphosphonate
Synonyms: bis(2-methylphenyl) methylphosphonate
Molecular Weight (g/mol): 276.27
Molecular Formula: C15H17O3P
InChi Key: BMXNSAMZRSPALK-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1OP(C)(=O)OC1=CC=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results