Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di-o-tolyl Methylphosphonate 98.0+%, TCI America™
SDP

Catalog No. D29891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D29891G Supplier TCI America Supplier No. D29891G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60146-72-7
Molecular Formula C15H17O3P
Molecular Weight (g/mol) 276.272
InChI Key BMXNSAMZRSPALK-UHFFFAOYSA-N
Synonym Methylphosphonic Acid Di-o-tolyl Ester
PubChem CID 185966
IUPAC Name 1-methyl-2-[methyl-(2-methylphenoxy)phosphoryl]oxybenzene
SMILES CC1=CC=CC=C1OP(=O)(C)OC2=CC=CC=C2C

Specifications

Color White
Quantity 1 g
Formula Weight 276.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Di-o-tolyl Methylphosphonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.