accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClNNCl

CAS RN 60168-88-9

IUPAC Name: (2-chlorophenyl)(4-chlorophenyl)(pyrimidin-5-yl)methanol
Synonyms: BLOC rimidin epa pesticide chemical code 206600 compound 56722 fenarimol rubigan 12 rc rubigan 12 RC caswell no. 207AA fenarimol (iso) rubigan-4
Molecular Weight (g/mol): 331.2
Molecular Formula: C17H12Cl2N2O
InChi Key: NHOWDZOIZKMVAI-UHFFFAOYNA-N
SMILES: OC(C1=CC=C(Cl)C=C1)(C1=CN=CN=C1)C1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results