CAS RN 602-56-2
IUPAC Name:
9-phenylacridine
Synonyms:
9-phenyl-acridine
Molecular Weight (g/mol):
255.32
Molecular Formula:
C19H13N
InChi Key:
MTRFEWTWIPAXLG-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=C2C=CC=CC2=NC2=CC=CC=C12
1
–
2
of
2
results
2
9-Phenylacridine 98.0+%, TCI America™
CAS: 602-56-2 Molecular Formula: C19H13N Molecular Weight (g/mol): 255.32 MDL Number: MFCD00092562 InChI Key: MTRFEWTWIPAXLG-UHFFFAOYSA-N PubChem CID: 69041 IUPAC Name: 9-phenylacridine SMILES: C1=CC=C(C=C1)C2=C3C=CC=CC3=NC4=CC=CC=C42