Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Phenylacridine, Thermo Scientific™

Catalog No. AC469430250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469430250 25 g
AC469430010 1 g
AC469430050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC469430250 Supplier Thermo Scientific Chemicals Supplier No. 469430250
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Alpha Vector PHENYLACRIDINE
Chemical Name or Material 9-Phenylacridine
CAS 602-56-2
Melting Point 183°C to 187°C
Color Yellow
Boiling Point 411°C
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C19 H13 N
Quantity 25 g
MDL Number 00092562
Molecular Weight (g/mol) 255.32
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.