accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOBr

CAS RN 602-77-7

IUPAC Name: 1,5-dibromo-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,5-dibromoanthracene-9,10-dione
Molecular Weight (g/mol): 366.01
Molecular Formula: C14H6Br2O2
InChi Key: SRRLOAAMZYZYQY-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C(=O)C1=C(C(Br)=CC=C1)C2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results