Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dibromoanthraquinone 93.0+%, TCI America™
SDP

Catalog No. D31811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D31811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D31811G Supplier TCI America Supplier No. D31811G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 602-77-7
Molecular Formula C14H6Br2O2
Molecular Weight (g/mol) 366.008
InChI Key SRRLOAAMZYZYQY-UHFFFAOYSA-N
PubChem CID 639109
IUPAC Name 1,5-dibromoanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Br)C(=O)C3=C(C2=O)C(=CC=C3)Br

Specifications

Melting Point 292°C
Quantity 1 g
Formula Weight 366.01
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dibromoanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.