accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONO

CAS RN 602-87-9

IUPAC Name: 5-nitro-1,2-dihydroacenaphthylene
Synonyms: 5-nitroacenaphthene
Molecular Weight (g/mol): 199.21
Molecular Formula: C12H9NO2
InChi Key: CUARLQDWYSRQDF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)