Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroacenaphthene, 98%, Thermo Scientific™

Catalog No. AAH5670303
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5670303 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5670303 Supplier Thermo Scientific Chemicals Supplier No. H5670303

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 602-87-9
Molecular Formula C12H9NO2
Quantity 1g
MDL Number MFCD00014277
Synonym 5-nitroacenaphthene, 5-nitroacenapthene, acenaphthene, 5-nitro, acenaphthylene, 1,2-dihydro-5-nitro, 5-nan, 1,2-dihydro-5-nitro-acenaphthylene, 5-nitronaphthalene ethylene, 5-nitronaphthalene, 1,2-dihydro-5-nitroacenaphthylene, ccris 438
InChI Key CUARLQDWYSRQDF-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23
IUPAC Name 5-nitro-1,2-dihydroacenaphthylene
Molecular Weight (g/mol) 199.21
PubChem CID 11769
ChEBI CHEBI:82350
Formula Weight 199.21
Percent Purity 98%
Chemical Name or Material 5-Nitroacenaphthene
Show More Show Less
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 210-025-0
RTECSNumber AB1060000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.