accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClClSH2NOOSNH2OO

CAS RN 60200-06-8

IUPAC Name: 4-amino-6-(1,2,2-trichloroethenyl)benzene-1,3-disulfonamide
Synonyms: clorsulon curatrem C8H8cl3N3O4S2 clorsulone clorsulonum
Molecular Weight (g/mol): 380.64
Molecular Formula: C8H8Cl3N3O4S2
InChi Key: QOVTVIYTBRHADL-UHFFFAOYSA-N
SMILES: NC1=CC(C(Cl)=C(Cl)Cl)=C(C=C1S(N)(=O)=O)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)