Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clorsulon, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18601761 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-601-761 5 kg
18-601-760 100 g
18-601-762 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-761 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C45335KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C4533, 60200-06-8, C8H8Cl3N3O4S2

Spectrum™ Chemical Clorsulon, USP is anthelmintic agent and antibiotic. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60200-06-8
Molecular Formula C8H8Cl3N3O4S2
Molecular Weight (g/mol) 380.64
InChI Key QOVTVIYTBRHADL-UHFFFAOYSA-N
IUPAC Name 4-amino-6-(1,2,2-trichloroethenyl)benzene-1,3-disulfonamide
SMILES NC1=CC(C(Cl)=C(Cl)Cl)=C(C=C1S(N)(=O)=O)S(N)(=O)=O

Specifications

Physical Form Powder
CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Sterility Non-sterile
Color White
Quantity 5 kg
Percent Purity 98 to 101%
Grade USP
Chemical Name or Material Clorsulon
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.