accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 60212-30-8

IUPAC Name: undec-3-yne
Synonyms: undec-3-yne
Molecular Weight (g/mol): 152.28
Molecular Formula: C11H20
InChi Key: DPWGJNPCPLQVKQ-UHFFFAOYSA-N
SMILES: CCCCCCCC#CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results