Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Undecyne 96.0+%, TCI America™
SDP

Catalog No. U00345ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00345ML Supplier TCI America Supplier No. U00345ML

Chemical Identifiers

CAS 60212-30-8
Molecular Formula C11H20
Molecular Weight (g/mol) 152.281
MDL Number MFCD00041663
InChI Key DPWGJNPCPLQVKQ-UHFFFAOYSA-N
Synonym 3-undecyne, acmc-209mii, dpwgjnpcplqvkq-uhfffaoysa
PubChem CID 143689
IUPAC Name undec-3-yne
SMILES CCCCCCCC#CCC

Specifications

Color Colorless
Boiling Point 80°C
Quantity 5 mL
Formula Weight 152.28
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Undecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.