accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3OCH3OOH

CAS RN 60241-74-9

IUPAC Name: 2,3,6-trimethoxybenzoic acid
Synonyms: 2,3,6-trimethoxybenzoic acid
Molecular Weight (g/mol): 212.2
Molecular Formula: C10H12O5
InChi Key: NSKSXYUOXAQHCH-UHFFFAOYSA-N
SMILES: COC1=C(OC)C(C(O)=O)=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results