accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3HNSiH3CCH3NHCH3H3C

CAS RN 6026-42-2

IUPAC Name: {dimethyl[(propan-2-yl)amino]silyl}(propan-2-yl)amine
Synonyms: isopropyl[(isopropylamino)dimethylsilyl]amine
Molecular Weight (g/mol): 174.36
Molecular Formula: C8H22N2Si
InChi Key: UNOQITWAUFOMKI-UHFFFAOYSA-N
SMILES: CC(C)N[Si](C)(C)NC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results