Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Di(isopropylamino)dimethylsilane, 95%, Thermo Scientific™

Catalog No. AC336630050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC336630050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336630050 Supplier Thermo Scientific Chemicals Supplier No. 336630050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6026-42-2
Molecular Formula C8H22N2Si
Molecular Weight (g/mol) 174.363
MDL Number MFCD01861707
InChI Key UNOQITWAUFOMKI-UHFFFAOYSA-N
Synonym 1,1-dimethyl-n,n'-diisopropylsilanediamine, di isopropylamino dimethylsilane, silanediamine, 1,1-dimethyl-n,n'-bis 1-methylethyl, bis isopropylamino dimethylsilane, di isopropylamino dimethyl-silane, dimethylsilanediylbis isopropylamine, di-isopropylamino dimethylsilane, n,n'-diisopropyl-1,1-dimethylsilanediamine, n-isopropylamino-dimethyl-silyl propan-2-amine, isopropyl isopropylamino dimethylsilyl amine
PubChem CID 80124
IUPAC Name N-[dimethyl-(propan-2-ylamino)silyl]propan-2-amine
SMILES CC(C)N[Si](C)(C)NC(C)C

Specifications

Color Colorless
Density 0.7983g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Refractive Index 1.4110 to 1.4150 (20°C, 589nm)
Linear Formula [(CH3)CH]Si(CH3)
Quantity 5 g
Formula Weight 174.36
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Di(isopropylamino)dimethylsilane, 95%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.