accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 60261-46-3

IUPAC Name: (2S)-2-[(diphenylphosphanyl)methyl]pyrrolidine
Synonyms: (2S)-2-[(diphenylphosphanyl)methyl]pyrrolidine
Molecular Weight (g/mol): 269.33
Molecular Formula: C17H20NP
InChi Key: IWRBGJKCDORZHK-GGYSOQFKNA-N
SMILES: C([C@@H]1CCCN1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)