Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-((Diphenylphosphino)methyl)pyrrolidine, 97%, Thermo Scientific™

Catalog No. AC434102500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434102500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC434102500 Supplier Thermo Scientific Chemicals Supplier No. 434102500
  • $162.00
    $131.65 / Each of 1

    Save $30.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60261-46-3
Molecular Formula C17H20NP
Molecular Weight (g/mol) 269.33
MDL Number MFCD16621443
InChI Key IWRBGJKCDORZHK-GGYSOQFKNA-N
Synonym s-2-diphenylphosphino methyl pyrrolidine, 2s-2-diphenylphosphanyl methyl pyrrolidine, pyrrolidine, 2-diphenylphosphino methyl-, 2s, aminophosphane
PubChem CID 11272693
IUPAC Name diphenyl-[[(2S)-pyrrolidin-2-yl]methyl]phosphane
SMILES C([C@@H]1CCCN1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Tan
Density 1.0400g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 250 mg
Specific Gravity 1.04
Formula Weight 269.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-2-((Diphenylphosphino)methyl)pyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.