accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNH2OOOHOHOOOHOHOHOHNHOH3C

CAS RN 60280-58-2

IUPAC Name: 2-amino-3-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}butanoic acid
Synonyms: 2-amino-3-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}butanoic acid
Molecular Weight (g/mol): 484.46
Molecular Formula: C18H32N2O13
InChi Key: VWNOCGQJSBAAFO-UHFFFAOYNA-N
SMILES: CC(OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1NC(C)=O)C(N)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results