Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gal beta(1-3)GalNAc-alpha-Thr 97.0+%, TCI America™
SDP

Catalog No. G03405MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03405MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G03405MG Supplier TCI America Supplier No. G03405MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity glycosylated amino acid for use as a laboratory reagent.

Chemical Identifiers

CAS 60280-58-2
Molecular Formula C18H32N2O13
Molecular Weight (g/mol) 484.46
MDL Number MFCD02683396
InChI Key VWNOCGQJSBAAFO-UHFFFAOYNA-N
Synonym O-[2-Acetamido-2-deoxy-3-O-(beta-D-galactopyranosyl)-alpha-D-galactopyranosyl]-L-threonine
PubChem CID 126968962
IUPAC Name 2-amino-3-{[3-acetamido-5-hydroxy-6-(hydroxymethyl)-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxan-2-yl]oxy}butanoic acid
SMILES CC(OC1OC(CO)C(O)C(OC2OC(CO)C(O)C(O)C2O)C1NC(C)=O)C(N)C(O)=O

Specifications

Color White
Quantity 5 mg
Formula Weight 484.46
Percent Purity ≥97.0% (HPLC)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material Gal beta(1-3)GalNAc-alpha-Thr
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.