accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 603-44-1

IUPAC Name: 4-[bis(4-hydroxyphenyl)methyl]phenol
Synonyms: 4-[bis(4-hydroxyphenyl)methyl]phenol
Molecular Weight (g/mol): 292.33
Molecular Formula: C19H16O3
InChi Key: WFCQTAXSWSWIHS-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results