Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4',4″-Trihydroxytriphenylmethane 98.0+%, TCI America™
SDP

Catalog No. T148425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T148425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T148425G Supplier TCI America Supplier No. T148425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 603-44-1
Molecular Formula C19H16O3
Molecular Weight (g/mol) 292.33
MDL Number MFCD00191589
InChI Key WFCQTAXSWSWIHS-UHFFFAOYSA-N
Synonym 4,4',4-methanetriyltriphenol, leucoaurin, 4,4',4-trihydroxytriphenylmethane, leucoaurine, leucaurin, phenol, methylidynetris, 4-bis 4-hydroxyphenyl methyl phenol, 4,4',4-methylidynetriphenol, 4,4',4-methylidynetrisphenol, phenol, 4,4',4-methylidynetris
PubChem CID 69047
IUPAC Name 4-[bis(4-hydroxyphenyl)methyl]phenol
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

Specifications

Melting Point 247°C
Quantity 25 g
Formula Weight 292.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′,4′′-Trihydroxytriphenylmethane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.